missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(p-cymene)ruthenium(II) dimer, 98%

Catalog No. aal1912604
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1912604 2 g
AAL19126ME 500 mg
AAL1912609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1912604 Supplier Thermo Scientific Chemicals Supplier No. L1912604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52462-29-0 | C20H28Cl4Ru2 | 612.384 g/mol

Dichloro(p-cymene)ruthenium(II) dimer is used as hydrosilylation catalyst and also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloro(p-cymene)ruthenium(II) dimer is used as hydrosilylation catalyst and also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 52462-29-0
Molecular Formula C20H28Cl4Ru2
Molecular Weight (g/mol) 612.384
MDL Number MFCD00064793
InChI Key LAXRNWSASWOFOT-UHFFFAOYSA-J
Synonym dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer
PubChem CID 10908223
IUPAC Name dichlororuthenium;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl

Specifications

Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 612.4
Percent Purity 98%
Chemical Name or Material Dichloro(p-cymene)ruthenium(II) dimer
Hazard Category H302-H317-H318
Hazard Statement GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
EINECSNumber 435-530-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only