Learn More
Dichloro(p-cymene)ruthenium(II) dimer, 98%
CAS: 52462-29-0 | C20H28Cl4Ru2 | 612.384 g/mol
Supplier: Thermo Scientific Chemicals L1912609
| Quantity | 10 g |
|---|
Description
Dichloro(p-cymene)ruthenium(II) dimer is used as hydrosilylation catalyst and also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDichloro(p-cymene)ruthenium(II) dimer is used as hydrosilylation catalyst and also used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
| 52462-29-0 | |
| 612.384 | |
| LAXRNWSASWOFOT-UHFFFAOYSA-J | |
| 10908223 | |
| CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl |
| C20H28Cl4Ru2 | |
| MFCD00064793 | |
| dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer | |
| dichlororuthenium;1-methyl-4-propan-2-ylbenzene |
Specifications
| Dichloro(p-cymene)ruthenium(II) dimer | |
| C20H28Cl4Ru2 | |
| MFCD00064793 | |
| Insoluble in water. | |
| CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)C(C)C.Cl[Ru]Cl.Cl[Ru]Cl | |
| 612.384 | |
| 612.4 |
| 52462-29-0 | |
| 10 g | |
| dichloro p-cymene ruthenium ii dimer, di-mu-chloro-bis chloro p-cymene ruthenium ii, rucl2 p-cymene 2, ru p-cymene cl2 2, dichloro p-cymene ruthenium dimer, p-cymene ruthenium ii chloride dimer, dichloro-p-cymene ruthenium ii dimer | |
| LAXRNWSASWOFOT-UHFFFAOYSA-J | |
| dichlororuthenium;1-methyl-4-propan-2-ylbenzene | |
| 10908223 | |
| 98% |
Safety and Handling
GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P501c
H302-H317-H318
EINECSNumber : 435-530-5
TSCA : No
Recommended Storage : Ambient temperatures