missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro(methyl)phenylsilane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 205°C |
| Quantity | 25 g |
| UN Number | 2437 |
| Formula Weight | 191.13 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dichloro(methyl)phenylsilane |
Safety and Handling
| EINECSNumber | (3)-2641 |
| RTECSNumber | VV3530000 |
| TSCA | Yes |