missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(methyl)phenylsilane 98.0+%, TCI America™

Catalog No. p-7134767
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P079025G 25 g
P0790250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P079025G Supplier TCI America Supplier No. P079025G
Only null left

Chemical Identifiers

CAS 149-74-6
Molecular Formula C7H8Cl2Si
Molecular Weight (g/mol) 191.13
MDL Number MFCD00000490
InChI Key GNEPOXWQWFSSOU-UHFFFAOYSA-N
Synonym methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
PubChem CID 9006
IUPAC Name dichloro(methyl)phenylsilane
SMILES C[Si](Cl)(Cl)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 205°C
Quantity 25 g
UN Number 2437
Formula Weight 191.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloro(methyl)phenylsilane
EINECSNumber (3)-2641
RTECSNumber VV3530000
TSCA Yes