missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(methyl)(3,3,3-trifluoropropyl)silane 98.0+%, TCI America™

Catalog No. p-7128428
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47295G 5 g
D472925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47295G Supplier TCI America Supplier No. D47295G

Chemical Identifiers

CAS 675-62-7
Molecular Formula C4H7Cl2F3Si
Molecular Weight (g/mol) 211.08
MDL Number MFCD00039294
InChI Key OHABWQNEJUUFAV-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropyl methyldichlorosilane, 3,3,3-trifluoropropyl dichloromethylsilane, dichloro methyl 3,3,3-trifluoropropyl silane, unii-38xgw8m24k, silane, dichloromethyl 3,3,3-trifluoropropyl, dichloromethyl-3,3,3-trifluoropropylsilane, dichloromethyl 3,3,3-trifluoropropyl silane, dichloro-methyl-3,3,3-trifluoropropyl silane, 4-04-00-04169 beilstein handbook reference, ksc491g7h
PubChem CID 12666
IUPAC Name dichloro-methyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](CCC(F)(F)F)(Cl)Cl

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
UN Number 2985
Formula Weight 211.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloro(methyl)(3,3,3-trifluoropropyl)silane
EINECSNumber (2)-2047
RTECSNumber VV3531500
TSCA Yes