missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(dimethylamino)phosphine 98.0+%, TCI America™

Supplier:  TCI America D32455G

Catalog No. D32455G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dichloro(dimethylamino)phosphine
Yellow
C2H6Cl2NP
2924
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
N-dichlorophosphanyl-N-methylmethanamine
136483
≥98.0% (GC)
683-85-2
150°C
5 g
dimethylphosphoramidous dichloride, dichloro dimethylamino phosphine, phosphoramidous dichloride, dimethyl, dimethylamino dichlorophosphine, dimethylphosphoramidousdichloride, me2npcl2, dimethylaminodichlorphosphine, dimethylaminodichlorophosphine, ch3 2npcl2, dichlorophosphanyl dimethylamine
CN(C)P(Cl)Cl
145.951
145.95
Liquid

Chemical Identifiers

683-85-2
145.951
dimethylphosphoramidous dichloride, dichloro dimethylamino phosphine, phosphoramidous dichloride, dimethyl, dimethylamino dichlorophosphine, dimethylphosphoramidousdichloride, me2npcl2, dimethylaminodichlorphosphine, dimethylaminodichlorophosphine, ch3 2npcl2, dichlorophosphanyl dimethylamine
N-dichlorophosphanyl-N-methylmethanamine
C2H6Cl2NP
XPWWDZRSNFSLRQ-UHFFFAOYSA-N
136483
CN(C)P(Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions