missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(diethylamino)phosphine 98.0+%, TCI America™

Catalog No. p-7127639
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32445G 5 g
D324425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32445G Supplier TCI America Supplier No. D32445G
Only null left

Chemical Identifiers

CAS 1069-08-5
Molecular Formula C4H10Cl2NP
Molecular Weight (g/mol) 174.005
InChI Key BPEMCEULJQTJMI-UHFFFAOYSA-N
Synonym diethylphosphoramidous dichloride, dichloro diethylamino phosphine, diethylaminodichlorophosphine, et2npcl2, acmc-2098mq, diethylamido dichlorophosphite, diethylamino dichlorophosphine, dichloro diethylamido phosphine, dichlorophosphanyl diethylamine, dichlorophosphorous diethylamide
PubChem CID 2733289
IUPAC Name N-dichlorophosphanyl-N-ethylethanamine
SMILES CCN(CC)P(Cl)Cl

Specifications

Color Yellow
Boiling Point 189°C
Quantity 5 g
UN Number 3265
Formula Weight 174.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloro(diethylamino)phosphine
TSCA No
Recommended Storage Refrigerator