missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)platinum(II), Pt 51.6-52.6%

Catalog No. p-7049079
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1273901 0.25 g
AA1273903 1 g
AA1273906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1273901 Supplier Thermo Scientific Chemicals Supplier No. 01273901

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12080-32-9 | C8H12Cl2Pt | 374.17 g/mol

it is used in Hydrosilyation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12080-32-9
Molecular Formula C8H12Cl2Pt
Molecular Weight (g/mol) 374.17
MDL Number MFCD00012413
InChI Key VVAOPCKKNIUEEU-PHFPKPIQSA-L
Synonym dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
PubChem CID 6436378
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
SMILES Cl[Pt]Cl.C1C\C=C/CC\C=C/1

Specifications

Melting Point 250°C (decomposition)
Quantity 0.25 g
Sensitivity Hygroscopic
Solubility Information Soluble in dichloromethane
Formula Weight 374.18
Concentration or Composition (by Analyte or Components) Pt 51.6 to 52.6%
Physical Form Solid
Chemical Name or Material Dichloro(1,5-cyclooctadiene)platinum(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-144-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only