missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)platinum(II), 98%, Thermo Scientific™

Chemical Identifiers

CAS 12080-32-9
Molecular Formula C8H12Cl2Pt
Molecular Weight (g/mol) 374.17
MDL Number MFCD00012413,MFCD00012413
InChI Key VVAOPCKKNIUEEU-PHFPKPIQSA-L
Synonym dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
PubChem CID 6436378
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
SMILES [Cl-].[Cl-].[Pt++].C1C\C=C/CC\C=C/1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC363612500
SDS View Documents
Thermo Scientific Chemicals
363612500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC363610010
SDS View Documents
Thermo Scientific Chemicals
363610010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12080-32-9
374.17
VVAOPCKKNIUEEU-PHFPKPIQSA-L
6436378
[Cl-].[Cl-].[Pt++].C1C\C=C/CC\C=C/1
C8H12Cl2Pt
MFCD00012413,MFCD00012413
dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride

Specifications

12080-32-9
97.5% min. (ex Pt)
C8H12Cl2Pt
250mg
Solubility in water: insoluble in water
[Cl-].[Cl-].[Pt++].C1C\C=C/CC\C=C/1
374.17
374.18
Dichloro(1, 5-cyclooctadiene)platinum(II)
Authentic
Glass bottle
MFCD00012413,MFCD00012413
dichloro 1,5-cyclooctadiene platinum ii, 1,5-cyclooctadienedichloroplatinum, dichloro 1,5-cyclooctadiene platinum, platinum, dichloro 1,5-cyclooctadiene, 1z,5z-cycloocta-1,5-diene; platinum 2+ ; dichloride, platinum, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, 1,5-cyclooctadieneplatinum ii dichloride, dichloro 1,5-cyclooctadiene platinum ii 250mg, dichloro 1,2,5,6-eta-cycloocta-1,5-diene platinum, platinum 2+ 1,5-cyclooctadiene, z,z-dichloride
VVAOPCKKNIUEEU-PHFPKPIQSA-L
(1Z,5Z)-cycloocta-1,5-diene;platinum(2+);dichloride
6436378
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 235-144-5

SDS
Documents

Documents

RUO – Research Use Only