missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro(1,10-phenanthroline)copper(II) 98.0+%, TCI America™
$82.02 - $242.33
Chemical Identifiers
| CAS | 14783-09-6 |
|---|---|
| Molecular Formula | C12H8Cl2CuN2 |
| Molecular Weight (g/mol) | 314.656 |
| MDL Number | MFCD00134224 |
| InChI Key | PHECBGBJCLDCMF-UHFFFAOYSA-L |
| Synonym | (1,10-Phenanthroline)copper(II) Dichloride |
| PubChem CID | 6100472 |
| IUPAC Name | dichlorocopper;1,10-phenanthroline |
| SMILES | C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.Cl[Cu]Cl |
Chemical Identifiers
| 14783-09-6 | |
| 314.656 | |
| PHECBGBJCLDCMF-UHFFFAOYSA-L | |
| 6100472 | |
| C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.Cl[Cu]Cl |
| C12H8Cl2CuN2 | |
| MFCD00134224 | |
| (1,10-Phenanthroline)copper(II) Dichloride | |
| dichlorocopper;1,10-phenanthroline |
Specifications
| 14783-09-6 | |
| C12H8Cl2CuN2 | |
| 1 g | |
| PHECBGBJCLDCMF-UHFFFAOYSA-L | |
| dichlorocopper;1,10-phenanthroline | |
| 6100472 | |
| ≥98.0% (N,T) | |
| Dichloro(1,10-phenanthroline)copper(II) |
| Green-Yellow | |
| MFCD00134224 | |
| (1,10-Phenanthroline)copper(II) Dichloride | |
| C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.Cl[Cu]Cl | |
| 314.656 | |
| 314.66 | |
| Crystalline Powder |
Safety and Handling
TSCA : No