missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Dic

Catalog No. D125407500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D125407500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. D125407500G Supplier Sigma Aldrich Supplier No. D125407500G
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DIC is an abbreviated form of N,N'sa-diisopropylcarbodiimide. It plays a role in the cyclization of N-(+a-hydroxy)amides to form 2-oxazolines.DIC (N,N'sa-Diisopropylcarbodiimide) has been used in combination with 1-hydroxy-7-azabenzotriazole (HOAt) for the coupling of amino acid with N-allylglycine to form N-allylpeptide.

Specifications

CAS 693-13-0
Density 0.815 g/mL at 20°C
Boiling Point 145°C to 148°C
Molecular Formula C7H14N2
Refractive Index n20/D 1.433
Linear Formula (CH3)2CHN=C=NCH(CH3)2
MDL Number MFCD00065689
Quantity 500 g
Synonym N,N'-Diisopropylcarbodiimide
Molecular Weight (g/mol) 126.2 g/mol