missing translation for 'onlineSavingsMsg'
Learn More

Dibutylboron trifluoromethanesulfonate, 1M sol. in dichloromethane

Catalog No. AC378990250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378990250 25 mL Glass Bottle
AC378991000 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC378990250 Supplier Thermo Scientific Chemicals Supplier No. 378990250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60669-69-4 | C9H18BF3O3S | 274.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Specifications

Chemical Name or Material Dibutylboron trifluoromethanesulfonate
CAS 60669-69-4, 75-09-2
Color Orange to Yellow
Density 1.2710g/mL
Flash Point 27°C
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C9H18BF3O3S
Linear Formula CF3SO3B[(CH2)3CH3]2
Quantity 25 mL
MDL Number MFCD00009669
Fieser 07,91; 09,140; 16,109
Specific Gravity 1.271
Synonym dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane
Solubility Information Solubility in water: reacts.
InChI Key FAVAVMFXAKZTMV-UHFFFAOYSA-N
SMILES CCCCB(CCCC)OS(=O)(=O)C(F)(F)F
IUPAC Name dibutylboranyl trifluoromethanesulfonate
Molecular Weight (g/mol) 274.11
PubChem CID 2724243
Formula Weight 274.11
Physical Form Solution
Show More Show Less

RUO – Research Use Only