missing translation for 'onlineSavingsMsg'
Learn More

Dibutylamine 99.0+%, TCI America™

Catalog No. p-7126090
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D021825ML 25 mL
D0218500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D021825ML Supplier TCI America Supplier No. D021825ML
Only null left

Chemical Identifiers

CAS 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point -59°C
Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 2248
Formula Weight 129.25
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Dibutylamine
EINECSNumber (2)-0137
RTECSNumber HR7780000
TSCA Yes