missing translation for 'onlineSavingsMsg'
Learn More

Dibutylamine (Reagent), Fisher Chemical™

$635.17 - $635.17

Chemical Identifiers

CAS 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
O2199-1
SDS View Documents
Fisher Chemical
O21991
1 L Glass Bottle
Each for $635.17
Case of 6 Each for $3,811.02
Only null left
 
Description

Description

Specifications

Chemical Identifiers

111-92-2
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
CCCCNCCCC
C8H19N
MFCD00009429
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
N-butylbutan-1-amine

Specifications

111-92-2
-62°C
159.6°C ± 0.1°C
Glass Bottle
1.4165 to 1.4185 (at 20°C)
MFCD00009429
2248
JQVDAXLFBXTEQA-UHFFFAOYSA-N
N-butylbutan-1-amine
4.46g/mol
129.25
Reagent
Dibutylamine
154.5° to 162.0°C
0.76g/cm³
47°C
C8H19N
[CH3(CH2)3]2NH
1 L
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
CCCCNCCCC
129.247
8148
2.59mmHg at 25°C
Liquid
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS
missing translation for 'documents'

missing translation for 'documents'