missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Dibutylamine

Catalog No. 4712321L
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4712321L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4712321L Supplier Sigma Aldrich Supplier No. 4712321L
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutylamine

Specifications

CAS 111-92-2
Melting Point -62°C (lit.)
Density 0.767 g/mL (at 25°C (literature))
Boiling Point 159°C (lit.)
Molecular Formula C8H19N
Refractive Index n20/D 1.417 (literature)
Linear Formula (CH3CH2CH2CH2)2 NH
MDL Number MFCD00009429
Quantity 1 L
Molecular Weight (g/mol) 129.24
Percent Purity ≥99.5%
Show More Show Less