missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Vinylboronate (stabilized with Phenothiazine) 94.0+%, TCI America™

Catalog No. D35965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35965G 5 g
D35961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35965G Supplier TCI America Supplier No. D35965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6336-45-4
Molecular Formula C10H21BO2
Molecular Weight (g/mol) 184.086
MDL Number MFCD00050049
InChI Key JQKJEPJLCSCBGW-UHFFFAOYSA-N
Synonym Vinylboronic Acid Dibutyl Ester
PubChem CID 236543
IUPAC Name dibutoxy(ethenyl)borane
SMILES B(C=C)(OCCCC)OCCCC

Specifications

Color Yellow
Boiling Point 89°C
Quantity 5 g
Formula Weight 184.09
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Vinylboronate (stabilized with Phenothiazine)
TSCA No
Recommended Storage Refrigerator