missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Sulfate 96.0+%, TCI America™

Supplier:  TCI America D163325G

 View more versions of this product

Catalog No. D163325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dibutyl Sulfate [Alkylating Agent]
Yellow
C8H18O4S
25 g
dibutylsulfate, di-n-butylsulfat, sulfuric acid, dibutyl ester, di-n-butylsulfate, di-n-butyl sulfate, dibutyl sulphate, di-n-butylsulfat german, sulfuric acid dibutyl ester, sulphuric acid dibutyl ester, dibutylsulfat
CCCCOS(=O)(=O)OCCCC
210.288
210.29
Liquid
625-22-9
141°C
MFCD00059420
2810
LMEDOLJKVASKTP-UHFFFAOYSA-N
dibutyl sulfate
12239
≥96.0% (GC)

Chemical Identifiers

625-22-9
210.288
LMEDOLJKVASKTP-UHFFFAOYSA-N
12239
CCCCOS(=O)(=O)OCCCC
C8H18O4S
MFCD00059420
dibutylsulfate, di-n-butylsulfat, sulfuric acid, dibutyl ester, di-n-butylsulfate, di-n-butyl sulfate, dibutyl sulphate, di-n-butylsulfat german, sulfuric acid dibutyl ester, sulphuric acid dibutyl ester, dibutylsulfat
dibutyl sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1678

RTECSNumber : WS7700000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions