missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Sulfate 96.0+%, TCI America™

Catalog No. D1633500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1633500G 500 g
D163325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1633500G Supplier TCI America Supplier No. D1633500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-22-9
Molecular Formula C8H18O4S
Molecular Weight (g/mol) 210.288
MDL Number MFCD00059420
InChI Key LMEDOLJKVASKTP-UHFFFAOYSA-N
Synonym dibutylsulfate, di-n-butylsulfat, sulfuric acid, dibutyl ester, di-n-butylsulfate, di-n-butyl sulfate, dibutyl sulphate, di-n-butylsulfat german, sulfuric acid dibutyl ester, sulphuric acid dibutyl ester, dibutylsulfat
PubChem CID 12239
IUPAC Name dibutyl sulfate
SMILES CCCCOS(=O)(=O)OCCCC

Specifications

Color Yellow
Boiling Point 141°C
Quantity 500 g
UN Number 2810
Formula Weight 210.29
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Sulfate [Alkylating Agent]
EINECSNumber (2)-1678
RTECSNumber WS7700000
TSCA No