missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Sebacate 98.0+%, TCI America™

Catalog No. p-7135768
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S002325ML 25 mL
S0023500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S002325ML Supplier TCI America Supplier No. S002325ML
Only null left

Chemical Identifiers

CAS 109-43-3
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.466
MDL Number MFCD00027218
InChI Key PYGXAGIECVVIOZ-UHFFFAOYSA-N
Synonym dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs
PubChem CID 7986
IUPAC Name dibutyl decanedioate
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 345°C
Quantity 25 mL
Formula Weight 314.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Sebacate
EINECSNumber (2)-0879
RTECSNumber VS1150000
TSCA Yes