missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl sebacate, 93%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 406650025

Quantity 2.5kg
Packaging Glass bottle

 View more versions of this product

Catalog No. AC406650025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

109-43-3
314.46
PYGXAGIECVVIOZ-UHFFFAOYSA-N
7986
CCCCOC(=O)CCCCCCCCC(=O)OCCCC
C18H34O4
MFCD00027218
dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs
dibutyl decanedioate

Specifications

Dibutyl sebacate
0.9400g/mL
Authentic
Glass bottle
1.4400 to 1.442
2.5kg
0.94
PYGXAGIECVVIOZ-UHFFFAOYSA-N
dibutyl decanedioate
7986
93%
109-43-3
167°C
91% min. (GC)
C18H34O4
CH3(CH2)3OCO(CH2)8COO(CH2)3CH3
MFCD00027218
dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs
CCCCOC(=O)CCCCCCCCC(=O)OCCCC
314.46
314.46
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-672-5

SDS
Documents

Documents