missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Oxalate 99.0+%, TCI America™

Catalog No. O0077500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0077500ML 500 mL
O007725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0077500ML Supplier TCI America Supplier No. O0077500ML
Only null left

Chemical Identifiers

CAS 2050-60-4
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009442
InChI Key JKRZOJADNVOXPM-UHFFFAOYSA-N
Synonym Oxalic Acid Dibutyl Ester
PubChem CID 16306
IUPAC Name dibutyl oxalate
SMILES CCCCOC(=O)C(=O)OCCCC

Specifications

Melting Point -29°C
Color Colorless
Boiling Point 246°C
Quantity 500 mL
Formula Weight 202.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Oxalate
EINECSNumber (2)-3789&(2)-0924
TSCA Yes