missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl Oxalate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | -29°C |
| Color | Colorless |
| Boiling Point | 246°C |
| Quantity | 25 mL |
| Formula Weight | 202.25 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dibutyl Oxalate |
Safety and Handling
| EINECSNumber | (2)-3789&(2)-0924 |
| TSCA | Yes |