missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Maleate 95.0+%, TCI America™

Numéro de catalogue. M0009500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M0009500G 500 g
M000925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M0009500G Fournisseur TCI America Code fournisseur M0009500G
Il en reste null

Identifiants chimiques

CAS 105-76-0
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00009447,MFCD00065141
InChI Key JBSLOWBPDRZSMB-FPLPWBNLSA-N
Synonym dibutyl maleate, butyl maleate, staflex dbm, rc comonomer dbm, maleic acid, dibutyl ester, di-n-butyl maleate, 2-butenedioic acid z-, dibutyl ester, 2-butenedioic acid, dibutyl ester, maleic acid dibutyl ester, 2-butenedioic acid 2z-, dibutyl ester
PubChem CID 5271569
IUPAC Name 1,4-dibutyl (2Z)-but-2-enedioate
SMILES CCCCOC(=O)\C=C/C(=O)OCCCC

Spécifications

Melting Point -85°C
Color Colorless
Boiling Point 161°C
Quantity 500 g
Formula Weight 228.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Maleate
EINECSNumber (2)-1107&(2)-1142
RTECSNumber ON0875000
TSCA Yes