missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl DL-Malate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 246.30 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dibutyl DL-Malate |
Safety and Handling
| EINECSNumber | (2)-1444 |
| TSCA | Yes |