missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Adipate 99.0+%, TCI America™

Catalog No. p-7116989
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A065525ML 25 mL
A0655500ML 500 mL
2 options

Catalog No. A065525ML

Supplier: TCI America A065525ML

Only null left

Chemical Identifiers

CAS 105-99-7
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.36
MDL Number MFCD00009451
InChI Key XTJFFFGAUHQWII-UHFFFAOYSA-N
Synonym dibutyl adipate, butyl adipate, di-n-butyl adipate, hexanedioic acid, dibutyl ester, cetiol b, polycizer w 260, adipic acid, dibutyl ester, unitolate b, experimental tick repellent 3, di-butyl adipate
PubChem CID 7784
ChEBI CHEBI:34686
IUPAC Name 1,6-dibutyl hexanedioate
SMILES CCCCOC(=O)CCCCC(=O)OCCCC

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 305°C
Quantity 25 mL
UN Number 3082
Formula Weight 258.36
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV0900000
TSCA Yes