missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Adipate 99.0+%, TCI America™

Catalog No. a0655500ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0655500ML 500 mL
A065525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0655500ML Supplier TCI America Supplier No. A0655500ML
Only null left

Chemical Identifiers

CAS 105-99-7
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.36
MDL Number MFCD00009451
InChI Key XTJFFFGAUHQWII-UHFFFAOYSA-N
Synonym dibutyl adipate, butyl adipate, di-n-butyl adipate, hexanedioic acid, dibutyl ester, cetiol b, polycizer w 260, adipic acid, dibutyl ester, unitolate b, experimental tick repellent 3, di-butyl adipate
PubChem CID 7784
ChEBI CHEBI:34686
IUPAC Name 1,6-dibutyl hexanedioate
SMILES CCCCOC(=O)CCCCC(=O)OCCCC

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 305°C
Quantity 500 mL
UN Number 3082
Formula Weight 258.36
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Adipate
EINECSNumber (2)-0861&(2)-0879
RTECSNumber AV0900000
TSCA Yes