missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyldithiocarbamic acid zinc salt, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 14726-36-4 |
|---|---|
| Molecular Formula | C30H28N2S4Zn |
| Molecular Weight (g/mol) | 610.21 |
| InChI Key | AUMBZPPBWALQRO-UHFFFAOYSA-L |
| Synonym | zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc |
| PubChem CID | 84610 |
| IUPAC Name | zinc;N,N-dibenzylcarbamodithioate |
| SMILES | C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC406440250
|
Thermo Scientific Chemicals
406440250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC406441000
|
Thermo Scientific Chemicals
406441000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 14726-36-4 | |
| 610.21 | |
| zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc | |
| zinc;N,N-dibenzylcarbamodithioate |
| C30H28N2S4Zn | |
| AUMBZPPBWALQRO-UHFFFAOYSA-L | |
| 84610 | |
| C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2] |
Specifications
| 14726-36-4 | |
| 95% | |
| C30H28N2S4Zn | |
| zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc | |
| C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2] | |
| 610.21 | |
| 610.21 | |
| Dibenzyldithiocarbamic acid zinc salt |
| Authentic | |
| Glass bottle | |
| 25g | |
| AUMBZPPBWALQRO-UHFFFAOYSA-L | |
| zinc;N,N-dibenzylcarbamodithioate | |
| 84610 | |
| 95% |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 238-778-
RUO â Research Use Only