missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyldithiocarbamic acid zinc salt, 95%

Catalog No. AC406440250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406440250 25 g Glass bottle
AC406441000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406440250 Supplier Thermo Scientific Chemicals Supplier No. 406440250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14726-36-4 | C30H28N2S4Zn | 610.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14726-36-4
Molecular Formula C30H28N2S4Zn
Molecular Weight (g/mol) 610.21
InChI Key AUMBZPPBWALQRO-UHFFFAOYSA-L
Synonym zinc ii dibenzyldithiocarbamate, zinc dibenzyldithiocarbamate, dibenzyldithiocarbamic acid zinc salt, dibenzyldithiocarbamic acid, zinc salt, zinc n,n-dibenzylcarbamodithioate, zinc dibenzyl dithiocarbamate, zinc bis dibenzyldithiocarbamate, acmc-209cyb, ksc910q0r, bis dibenzyldithiocarbamic acid zinc
PubChem CID 84610
IUPAC Name zinc;N,N-dibenzylcarbamodithioate
SMILES C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].C1=CC=C(C=C1)CN(CC2=CC=CC=C2)C(=S)[S-].[Zn+2]

Specifications

Melting Point 183.0°C to 188.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Formula Weight 610.21
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Dibenzyldithiocarbamic acid zinc salt
Hazard Category GHS Signal Word: Warning
EINECSNumber 238-778-

RUO – Research Use Only