missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl 99.0+%, TCI America™

Catalog No. p-7118457
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
B045925G 25 g
B0459100G 100 g
B0459500G 500 g
3 options

Catalog No. B045925G

Supplier: TCI America B045925G

Only null left

Chemical Identifiers

CAS 103-29-7
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00004796
InChI Key QWUWMCYKGHVNAV-UHFFFAOYSA-N
Synonym 1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis
PubChem CID 7647
ChEBI CHEBI:34047
IUPAC Name (2-phenylethyl)benzene
SMILES C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 52°C
Color White
Boiling Point 284°C
Quantity 25 g
Formula Weight 182.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl
EINECSNumber (4)-1418
RTECSNumber DT4375000
TSCA Yes