missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl Malonate 95.0+%, TCI America™
Supplier: TCI America M1080100G
Specifications
| Dibenzyl Malonate | |
| Yellow | |
| C17H16O4 | |
| 100 g | |
| RYFCSKVXWRJEOB-UHFFFAOYSA-N | |
| dibenzyl propanedioate | |
| 84754 | |
| ≥95.0% (GC) |
| 15014-25-2 | |
| 234°C | |
| MFCD00004779 | |
| dibenzyl malonate, propanedioic acid, bis phenylmethyl ester, 1,3-dibenzyl propanedioate, malonic acid dibenzyl ester, malonic acid dibenzyl, acmc-209d3u, diphenylmethyl propane-1,3-dioate, propanedioic acid,1,3-bis phenylmethyl ester, propanedioic acid, 1,3-bis phenylmethyl ester, s-3-amino-2-oxetanone p-toluenesulfonic acid salt boc protection form | |
| C1=CC=C(C=C1)COC(=O)CC(=O)OCC2=CC=CC=C2 | |
| 284.311 | |
| 284.31 | |
| Liquid |
Chemical Identifiers
| 15014-25-2 | |
| 284.311 | |
| RYFCSKVXWRJEOB-UHFFFAOYSA-N | |
| dibenzyl propanedioate |
| C17H16O4 | |
| MFCD00004779 | |
| 84754 | |
| C1=CC=C(C=C1)COC(=O)CC(=O)OCC2=CC=CC=C2 |
Safety and Handling
TSCA : No