missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Malonate 95.0+%, TCI America™

Catalog No. M108025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M108025G 25 g
M1080100G 100 g
M1080500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M108025G Supplier TCI America Supplier No. M108025G

Chemical Identifiers

CAS 15014-25-2
Molecular Formula C17H16O4
Molecular Weight (g/mol) 284.311
MDL Number MFCD00004779
InChI Key RYFCSKVXWRJEOB-UHFFFAOYSA-N
PubChem CID 84754
IUPAC Name dibenzyl propanedioate
SMILES C1=CC=C(C=C1)COC(=O)CC(=O)OCC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 234°C
Quantity 25 g
Synonym dibenzyl malonate, propanedioic acid, bis phenylmethyl ester, 1,3-dibenzyl propanedioate, malonic acid dibenzyl ester, malonic acid dibenzyl, acmc-209d3u, diphenylmethyl propane-1,3-dioate, propanedioic acid,1,3-bis phenylmethyl ester, propanedioic acid, 1,3-bis phenylmethyl ester, s-3-amino-2-oxetanone p-toluenesulfonic acid salt boc protection form
Formula Weight 284.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibenzyl Malonate