missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl ketoxime, 98+%

Catalog No. p-7043470
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0091406 5 g
AAL0091414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0091406 Supplier Thermo Scientific Chemicals Supplier No. L0091406
Only null left

CAS: 1788-31-4 | C15H15NO | 225.291 g/mol

Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1788-31-4
Molecular Formula C15H15NO
Molecular Weight (g/mol) 225.291
MDL Number MFCD00015453
InChI Key SXEBLVKLMOIGER-UHFFFAOYSA-N
Synonym dibenzyl ketoxime, 1,3-diphenylacetone oxime, 1,3-diphenylpropan-2-one oxime, 2-propanone, 1,3-diphenyl-, oxime, n-1,3-diphenylpropan-2-ylidene hydroxylamine, 2-hydroxyimino-1,3-diphenylpropane, dibenzylketoxim, maybridge4_000477, dibenzyl ketoxime, 98+%, 1,3-diphenylacetone oxime #
PubChem CID 74518
IUPAC Name N-(1,3-diphenylpropan-2-ylidene)hydroxylamine
SMILES C1=CC=C(C=C1)CC(=NO)CC2=CC=CC=C2

Specifications

Melting Point 122°C to 124°C
Quantity 5 g
Beilstein 1965977
Solubility Information Insoluble in water.
Formula Weight 225.29
Percent Purity ≥98%
Chemical Name or Material Dibenzyl ketoxime
EINECSNumber 217-251-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only