missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Carbonate 98.0+%, TCI America™

Catalog No. C16001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16001G 1 g
C16005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16001G Supplier TCI America Supplier No. C16001G

Chemical Identifiers

CAS 3459-92-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00014436
InChI Key PIZLBWGMERQCOC-UHFFFAOYSA-N
Synonym carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester
PubChem CID 77002
IUPAC Name dibenzyl carbonate
SMILES O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Dibenzyl Carbonate