missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl carbonate, 98%

Catalog No. p-7081450
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1146409 10 g
AAA1146418 50 g
AAA1146430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1146409 Supplier Thermo Scientific Chemicals Supplier No. A1146409
Only null left

CAS: 3459-92-5 | C15H14O3 | 242.27 g/mol

Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ionic liquids can effectively accelerate slow N-benzylation reactions utilizing dibenzyl carbonate as an alkylating reagent. Dibenzyl carbonate (DBzlC) has been used to benzylate phenylacetonitrile, benzyl phenylacetate and phenol. Selective N, N-dibenzylation of primary aliphatic amines was carried with dibenzyl carbonate in the presence of phosphonium salts.

Solubility
Sparingly Soluble (0.099 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 3459-92-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00014436
InChI Key PIZLBWGMERQCOC-UHFFFAOYSA-N
Synonym carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester
PubChem CID 77002
IUPAC Name dibenzyl carbonate
SMILES O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Melting Point 27°C to 30°C
Boiling Point 150°C to 155°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5485
Quantity 10 g
Beilstein 1882864
Sensitivity Air sensitive
Solubility Information Sparingly Soluble (0.099g/L) (25°C).
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material Dibenzyl carbonate
Show More Show Less
Hazard Category H302+H312
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
EINECSNumber 222-401-1
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only