missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric Acid 98.0+%, TCI America™

Catalog No. d3492250g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
D3492250G 250 g
D349225G 25 g
2 options

Catalog No. D3492250G

Supplier: TCI America D3492250G

Only null left

Chemical Identifiers

CAS 2743-38-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00003074
InChI Key YONLFQNRGZXBBF-ZIAGYGMSSA-N
Synonym dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid
PubChem CID 75969
IUPAC Name (2R,3R)-2,3-bis(benzoyloxy)butanedioic acid
SMILES OC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 153°C
Color White
Boiling Point 234°C
Quantity 250 g
Formula Weight 358.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Dibenzoyl-L-tartaric Acid
EINECSNumber (3)-1290
RTECSNumber WW8070000
TSCA Yes