missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric Acid 98.0+%, TCI America™

Catalog No. p-7127942
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
D382625G 25 g
D3826250G 250 g
2 options

Catalog No. D382625G

Supplier: TCI America D382625G

Only null left

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
IUPAC Name (2S,3S)-2,3-bis(benzoyloxy)butanedioic acid
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C
Color White
Quantity 25 g
Formula Weight 358.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Dibenzoyl-D-tartaric Acid
EINECSNumber (3)-1290
TSCA Yes