missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzothiophene 98.0+%, TCI America™
$51.98 - $186.74
Chemical Identifiers
| CAS | 132-65-0 |
|---|---|
| Molecular Formula | C12H8S |
| Molecular Weight (g/mol) | 184.256 |
| MDL Number | MFCD00004969 |
| InChI Key | IYYZUPMFVPLQIF-UHFFFAOYSA-N |
| Synonym | diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene |
| PubChem CID | 3023 |
| ChEBI | CHEBI:23681 |
| IUPAC Name | dibenzothiophene |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3S2 |
Chemical Identifiers
| 132-65-0 | |
| 184.256 | |
| IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
| 3023 | |
| dibenzothiophene |
| C12H8S | |
| MFCD00004969 | |
| diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene | |
| CHEBI:23681 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3S2 |
Specifications
| 132-65-0 | |
| White-Yellow | |
| C12H8S | |
| 5 g | |
| diphenylene sulfide, dibenzo b,d thiophene, 9-thiafluorene, alpha-thiafluorene, 2,2'-biphenylylene sulfide, usan, unii-z3d4aj1r48, 1,1'-biphenyl-2,2'-diyl sulfide, .alpha.-thiafluorene | |
| C1=CC=C2C(=C1)C3=CC=CC=C3S2 | |
| 184.256 | |
| CHEBI:23681 | |
| ≥98.0% (GC) | |
| Dibenzothiophene |
| 99°C | |
| 333°C | |
| MFCD00004969 | |
| 3077 | |
| IYYZUPMFVPLQIF-UHFFFAOYSA-N | |
| dibenzothiophene | |
| 3023 | |
| 184.26 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3352
RTECSNumber : HQ3490550
TSCA : Yes