missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-5,5-dioxide, Thermo Scientific™

Catalog No. ac112550250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112550250 Supplier Thermo Scientific Chemicals Supplier No. 112550250
Only null left

Chemical Identifiers

CAS 1016-05-3
Molecular Formula C12H8O2S
Molecular Weight (g/mol) 216.25
MDL Number 00004970
InChI Key IKJFYINYNJYDTA-UHFFFAOYSA-N
IUPAC Name 8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(13),2,4,6,9,11-hexaene-8,8-dione
SMILES O=S1(=O)C2=CC=CC=C2C2=CC=CC=C12

Specifications

Alpha Vector DIBENZOTHIOPHENESULFONE
Melting Point 231°C to 236°C
Color White to yellow
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 25 g
Beilstein 17-72
Physical Form Crystalline Powder
Chemical Name or Material Dibenzothiophene-5,5-dioxide
EINECSNumber 213-805-9
Recommended Storage Normal conditions