missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene 5,5-Dioxide 98.0+%, TCI America™

Catalog No. d415325g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D415325G 25 g
D41535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D415325G Supplier TCI America Supplier No. D415325G
Only null left

Chemical Identifiers

CAS 1016-05-3
Molecular Formula C12H8O2S
Molecular Weight (g/mol) 216.254
MDL Number MFCD00004970
InChI Key IKJFYINYNJYDTA-UHFFFAOYSA-N
Synonym dibenzothiophene sulfone, dibenzo b,d thiophene 5,5-dioxide, dibenzothiophene-5,5-dioxide, dibenzothiophene, 5,5-dioxide, dibenzothiophene dioxide, diphenylene sulfone, dbto2, dibenzothiophene s,s-dioxide, dibenzosulfolane, dbt-sulfone
PubChem CID 13908
IUPAC Name dibenzothiophene 5,5-dioxide
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3S2(=O)=O

Specifications

Melting Point 236°C
Color White
Quantity 25 g
Formula Weight 216.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzothiophene 5,5-Dioxide