missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. D47671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47671G 1 g
D47675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47671G Supplier TCI America Supplier No. D47671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23985-81-1
Molecular Formula C13H8OS
Molecular Weight (g/mol) 212.266
MDL Number MFCD00961967
InChI Key XESZAOMRYLSHOM-UHFFFAOYSA-N
Synonym dibenzo b,d thiophene-4-carbaldehyde, dibenzothiophene-4-carboxaldehyde, dibenzo b,d thiophene-4-carboxaldehyde, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-6-carbaldehyde, 4-dibenzothiophenecarboxaldehyde, benzo b benzo b thiophene-4-carbaldehyde, dibenzo b,d ?thiophene-?4-?carbaldehyde, 8-thiatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene-6-carbaldehyde, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,5,9,11-hexaene-6-carbaldehyde
PubChem CID 641359
IUPAC Name dibenzothiophene-4-carbaldehyde
SMILES C1=CC=C2C(=C1)C3=C(S2)C(=CC=C3)C=O

Specifications

Melting Point 127°C
Color Yellow
Quantity 1 g
Formula Weight 212.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzothiophene-4-carboxaldehyde