missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzothiophene-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
$63.52 - $63.52
Chemical Identifiers
| CAS | 108847-20-7 |
|---|---|
| Molecular Formula | C12H9BO2S |
| Molecular Weight (g/mol) | 228.07 |
| MDL Number | MFCD01318182 |
| InChI Key | GOXNHPQCCUVWRO-UHFFFAOYSA-N |
| PubChem CID | 2734329 |
| IUPAC Name | {8-thiatricyclo[7.4.0.0²,â·]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid |
| SMILES | OB(O)C1=CC=CC2=C1SC1=C2C=CC=C1 |
Chemical Identifiers
| 108847-20-7 | |
| 228.07 | |
| GOXNHPQCCUVWRO-UHFFFAOYSA-N | |
| {8-thiatricyclo[7.4.0.0²,â·]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid |
| C12H9BO2S | |
| MFCD01318182 | |
| 2734329 | |
| OB(O)C1=CC=CC2=C1SC1=C2C=CC=C1 |
Specifications
| 108847-20-7 | |
| White | |
| MFCD01318182 | |
| dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid | |
| OB(O)C1=CC=CC2=C1SC1=C2C=CC=C1 | |
| 228.07 | |
| 228.07 | |
| Dibenzothiophene-4-boronic Acid (contains varying amounts of Anhydride) |
| 339°C | |
| C12H9BO2S | |
| 1 g | |
| GOXNHPQCCUVWRO-UHFFFAOYSA-N | |
| {8-thiatricyclo[7.4.0.0²,â·]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid | |
| 2734329 | |
| Crystalline Powder |
Safety and Handling
TSCA : No