missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-4-boronic acid, 95%

Catalog No. AAL1983106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1983106 5 g
AAL1983103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1983106 Supplier Thermo Scientific Chemicals Supplier No. L1983106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108847-20-7 | C12H9BO2S | 228.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108847-20-7
Molecular Formula C12H9BO2S
Molecular Weight (g/mol) 228.07
MDL Number MFCD01318182
InChI Key GOXNHPQCCUVWRO-UHFFFAOYSA-N
Synonym dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
PubChem CID 2734329
IUPAC Name dibenzothiophen-4-ylboronic acid
SMILES OB(O)C1=CC=CC2=C1SC1=C2C=CC=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 197863
Formula Weight 228.08
Percent Purity 95%
Chemical Name or Material Dibenzothiophene-4-boronic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only