missing translation for 'onlineSavingsMsg'
Learn More

Dibenzothiophene-4-boronic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 359080010

Quantity 1g

 View more versions of this product

Catalog No. AC359080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

108847-20-7
228.08
dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
dibenzothiophen-4-ylboronic acid
C12H9BO2S
GOXNHPQCCUVWRO-UHFFFAOYSA-N
2734329
B(C1=C2C(=CC=C1)C3=CC=CC=C3S2)(O)O

Specifications

Dibenzothiophene-4-boronic acid
Authentic
Glass Bottle
1g
GOXNHPQCCUVWRO-UHFFFAOYSA-N
dibenzothiophen-4-ylboronic acid
2734329
95%
108847-20-7
94% min. (HPLC)
C12H9BO2S
dibenzothiophene-4-boronic acid, 4-dibenzothiopheneboronic acid, dibenzo b,d thiophen-4-ylboronic acid, 4-dibenzothienylboronic acid, dibenzo b,d thiophene-4-boronic acid, boronic acid, 4-dibenzothienyl, 8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid, pubchem7386, 4-dibenzothiopheneboronicacid
B(C1=C2C(=CC=C1)C3=CC=CC=C3S2)(O)O
228.08
228.08
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions