missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzothiophene-2-carboxaldehyde 98.0+%, TCI America™
$533.13 - $1860.33
Chemical Identifiers
| CAS | 22099-23-6 |
|---|---|
| Molecular Formula | C13H8OS |
| Molecular Weight (g/mol) | 212.266 |
| MDL Number | MFCD02251670 |
| InChI Key | QFGXAEFJJINHAI-UHFFFAOYSA-N |
| PubChem CID | 641360 |
| IUPAC Name | dibenzothiophene-2-carbaldehyde |
| SMILES | C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)C=O |
Chemical Identifiers
| 22099-23-6 | |
| 212.266 | |
| QFGXAEFJJINHAI-UHFFFAOYSA-N | |
| dibenzothiophene-2-carbaldehyde |
| C13H8OS | |
| MFCD02251670 | |
| 641360 | |
| C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)C=O |
Specifications
| 22099-23-6 | |
| White-Yellow | |
| MFCD02251670 | |
| QFGXAEFJJINHAI-UHFFFAOYSA-N | |
| dibenzothiophene-2-carbaldehyde | |
| 641360 | |
| ≥98.0% (GC) | |
| Dibenzothiophene-2-carboxaldehyde |
| 107°C | |
| C13H8OS | |
| 1 g | |
| C1=CC=C2C(=C1)C3=C(S2)C=CC(=C3)C=O | |
| 212.266 | |
| 212.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No