missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. D47715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47715G 5 g
D47711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47715G Supplier TCI America Supplier No. D47715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96706-46-6
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.205
MDL Number MFCD03306027
InChI Key GQYTWBPRZFRASB-UHFFFAOYSA-N
PubChem CID 126043
IUPAC Name dibenzofuran-4-carbaldehyde
SMILES C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)C=O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzofuran-4-carboxaldehyde