missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-18-crown-6, 98+%

Catalog No. p-7020986
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1313306 5 g
AAA1313314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1313306 Supplier Thermo Scientific Chemicals Supplier No. A1313306
Only null left

CAS: 14187-32-7 | C20H24O6 | 360.406 g/mol

Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.

Chemical Identifiers

CAS 14187-32-7
Molecular Formula C20H24O6
MDL Number MFCD00005098
InChI Key YSSSPARMOAYJTE-UHFFFAOYSA-N
Synonym dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
PubChem CID 26541
ChEBI CHEBI:358732

Specifications

Melting Point 160°C to 164°C
Boiling Point 380°C to 384°C (679 mmHg)
Quantity 5 g
Beilstein 1162153
Merck Index 14,2602
Solubility Information Sparingly soluble in water.
SMILES C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
Molecular Weight (g/mol) 360.406
Formula Weight 360.41
Percent Purity ≥98%
Chemical Name or Material Dibenzo-18-crown-6
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-041-3
RTECSNumber HP5386000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only