missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-18-crown-6, 98+%

CAS: 14187-32-7 | C20H24O6 | 360.4 g/mol

$101.24 - $101.24

Chemical Identifiers

CAS 14187-32-7
Molecular Formula C20H24O6
Molecular Weight (g/mol) 360.4
MDL Number MFCD00005098
InChI Key YSSSPARMOAYJTE-UHFFFAOYSA-N
Synonym dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
PubChem CID 26541
ChEBI CHEBI:358732
SMILES C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169800100
SDS View Documents
Thermo Scientific Chemicals
169800100
10 g Glass bottle
Each for $101.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
Specifications

Chemical Identifiers

14187-32-7
360.4
YSSSPARMOAYJTE-UHFFFAOYSA-N
26541
C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
C20H24O6
MFCD00005098
dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
CHEBI:358732

Specifications

14187-32-7
White to Beige
Authentic
Glass bottle
MFCD00005098
06,159; 08,128; 09,126; 12,147
dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
YSSSPARMOAYJTE-UHFFFAOYSA-N
360.4
CHEBI:358732
98+%
Dibenzo-18-crown-6, 98%
160.0°C to 164.0°C
380.0°C to 384.0°C (679.0 mmHg)
98% min. (HPLC)
C20H24O6
10 g
14, 2602
Solubility in water: insoluble. Other solubilities: soluble in chloroform,toluene,ether,etc
C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1
26541
360.4
Fluffy Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 238-041-3

RTECSNumber : HP5386000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only