missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo-18-crown-6, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
Specifications
Specifications
| Melting Point | 160.0°C to 164.0°C |
| Color | White to Beige |
| Boiling Point | 380.0°C to 384.0°C (679.0 mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 50 g |
| Fieser | 06,159; 08,128; 09,126; 12,147 |
| Merck Index | 14, 2602 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in chloroform,toluene,ether,etc |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Avoid breathing dust/fume/ |
| EINECSNumber | 238-041-3 |
| RTECSNumber | HP5386000 |
| TSCA | TSCA |
RUO – Research Use Only