missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-18-crown-6, 98+%

Catalog No. AC169800100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169800100 10 g Glass bottle
AC169800500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169800100 Supplier Thermo Scientific Chemicals Supplier No. 169800100
Only null left

CAS: 14187-32-7 | C20H24O6 | 360.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 14187-32-7
Molecular Formula C20H24O6
Molecular Weight (g/mol) 360.4
MDL Number MFCD00005098
InChI Key YSSSPARMOAYJTE-UHFFFAOYSA-N
Synonym dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
PubChem CID 26541
ChEBI CHEBI:358732
SMILES C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1

Specifications

Melting Point 160.0°C to 164.0°C
Color White to Beige
Boiling Point 380.0°C to 384.0°C (679.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Fieser 06,159; 08,128; 09,126; 12,147
Merck Index 14, 2602
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,toluene,ether,etc
Formula Weight 360.4
Percent Purity 98+%
Physical Form Fluffy Powder
Chemical Name or Material Dibenzo-18-crown-6, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
EINECSNumber 238-041-3
RTECSNumber HP5386000
TSCA TSCA

RUO – Research Use Only