missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo-18-crown-6, 98+%
Description
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.
Dibenzo-18-crown-6, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is an important organic intermediate.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to strong oxidizing agents, strong acids.
Specifications
Specifications
| Melting Point | 160°C to 164°C |
| Boiling Point | 380°C to 384°C (679 mmHg) |
| Quantity | 25 g |
| Beilstein | 1162153 |
| Merck Index | 14,2602 |
| Solubility Information | Sparingly soluble in water. |
| SMILES | C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1 |
| Molecular Weight (g/mol) | 360.406 |
| Formula Weight | 360.41 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 238-041-3 |
| RTECSNumber | HP5386000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only